Skip to Content
MilliporeSigma

Skip To

1154503

USP

Cyclobenzaprine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-(3-Dimethylaminopropylidene)dibenzo[a,e]cycloheptatriene hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H21N · HCl
CAS Number:
Molecular Weight:
311.85
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

cyclobenzaprine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2C=Cc3ccccc13

InChI

1S/C20H21N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-14H,7,15H2,1-2H3;1H

InChI key

VXEAYBOGHINOKW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclobenzaprine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amitriptyline Hydrochloride
  • Cyclobenzaprine Hydrochloride
  • Cyclobenzaprine Hydrochloride Extended-Release Capsules
  • Cyclobenzaprine Hydrochloride Tablets

Biochem/physiol Actions

Skeletal muscle relaxant; 5-HT2 serotonin receptor antagonist.
Skeletal muscle relaxant; reduces muscle spasm by depression of brainstem neurons; 5-HT2 serotonin receptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C3200000BP886PHR2797
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical small molecule

API family

cyclobenzaprine

API family

cyclobenzaprine

API family

cyclobenzaprine

API family

cyclobenzaprine


Still not finding the right product?

Explore all of our products under Cyclobenzaprine hydrochloride


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1154503-200MG04061838703279

Questions

Reviews

No rating value

Active Filters