Skip to Content
MilliporeSigma

Skip To

1111908

USP

p-Chloroaniline

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Chloroaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
471359
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor density

4.4 (vs air)

vapor pressure

0.15 mmHg ( 25 °C)

API family

chlorhexidine , proguanil

manufacturer/tradename

USP

bp

232 °C (lit.)

mp

67-70 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(Cl)cc1

InChI

1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

QSNSCYSYFYORTR-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

p-Chloroaniline USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Chlorhexidine Gluconate Topical Gel
  • Chlorhexidine Acetate
  • Chlorhexidine Hydrochloride
  • Chlorhexidine Gluconate Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR12008.02613477222
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

format

neat

format

neat

format

-

format

-

API family

chlorhexidine , proguanil

API family

proguanil, chlorhexidine 

API family

-

API family

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-


Still not finding the right product?

Explore all of our products under p-Chloroaniline


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1111908-200MG04061838698599

Questions

Reviews

No rating value

Active Filters