Skip to Content
MilliporeSigma

Skip To

1076400

USP

Brimonidine tartrate

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H16BrN5O6
CAS Number:
Molecular Weight:
442.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

brimonidine

form

powder

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Brc1c2nccnc2ccc1NC3=NCCN3.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1

InChI key

QZHBYNSSDLTCRG-LREBCSMRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Brimonidine tartrate USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, system suitability, and sensitivity solutions during the assay, and impurity analysis of brimonidine tartrate.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0001658CDS022538PHR2795
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

form

powder

form

-

form

crystals

form

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

API family

brimonidine

API family

brimonidine

API family

-

API family

brimonidine


Still not finding the right product?

Explore all of our products under Brimonidine tartrate


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Brimonidine Tartrate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(4), 591-591 (2020)



Global Trade Item Number

SKUGTIN
1076400-250MG04061835345243

Questions

Reviews

No rating value

Active Filters