Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
product line
TraceCERT®
grade
certified reference material, TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
format
neat
storage temp.
2-8°C
SMILES string
FC(C(F)(F)F)C(F)(F)Oc1cc(Cl)c(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl
InChI
1S/C17H8Cl2F8N2O3/c18-6-5-11(32-17(26,27)14(22)16(23,24)25)7(19)4-10(6)28-15(31)29-13(30)12-8(20)2-1-3-9(12)21/h1-5,14H,(H2,28,29,30,31)
InChI key
PWPJGUXAGUPAHP-UHFFFAOYSA-N
General description
Lufenuron is an insect growth regulator used to control biting and sucking insects and belongs to the group of benzoylphenylurea insecticides. This systemic insecticide is known to be an inhibitor of chitin synthesis in the cuticle of insects. Lufenuron is used for the control of lepidopteran and coleopteran larvae and is extensively applied in cotton, maize, sugar beet, potatoes, grapes, citrus, and ornamentals farms. Lufenuron is also used in veterinary medicine as a flea control treatment and as an anti-fungal agent for dogs and cats.
Lufenuron is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009 (repealing Directive 91/414/EEC). However, the maximum residue level (MRL) for Lufenuron (any ratio of constituent isomers) is set from 0.01 mg/kg to 2 mg/kg for products of plant and animal origin following Reg. (EU) 2020/856. The Korea Food and Drug Administration (KFDA) has established an MRL of 0.01 to 0.2 ppm.
Lufenuron has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.
Application
- Evaluate the dissipation of lufenuron residues as a function of time and calculate the pre-harvest intervals on treated Chinese cabbages grown under greenhouse conditions
- Determine the effect of lufenuron exposure on different health biomarkers of common carp (Cyprinus carpio)
- Examine the effects of chitin-synthesis interruption during the development and reproduction of Drosophila melanogaster when administered with lufenuron
- Study the effects of lufenuron on the development and mortality of Lobesia botrana eggs and larvae and also on adult fecundity, fertility, and longevity
- Investigate the presence of chitin in ovaries of Rhodnius prolixus and study the effect of lufenuron on oogenesis and egg-laying
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format neat | format - | format neat | format neat |
| grade certified reference material, TraceCERT® | grade certified reference material, pharmaceutical secondary standard | grade analytical standard | grade pharmaceutical primary standard |
| manufacturer/tradename Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename USP |
| product line TraceCERT® | product line - | product line PESTANAL® | product line - |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life limited shelf life, expiry date on the label | shelf life - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under Lufenuron
flash_point_f
>338.0 °F - (External MSDS)
flash_point_c
> 170 °C - (External MSDS)
target_organs
Lungs,Liver,Central nervous system,Stomach
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1 - STOT RE 1 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



