Skip to Content
MilliporeSigma

Skip To

T4272

Trehalose 6-phosphate dipotassium salt

~95% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C12H21O14PK2
CAS Number:
Molecular Weight:
498.46
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95% (TLC)

Quality Level

form

powder

impurities

<10% water (Karl Fischer)

color

off-white,



cation traces

K: 14-17.3%, P: 5.8 - 6.7%

application(s)

agriculture

storage temp.

−20°C

SMILES string

[K].[H][C@@]1(O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)O[C@H]2O[C@H](COP(O)(O)=O)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C12H23O14P.K.H/c13-1-3-5(14)7(16)9(18)11(24-3)26-12-10(19)8(17)6(15)4(25-12)2-23-27(20,21)22;;/h3-19H,1-2H2,(H2,20,21,22);;/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

KSPIKJRSOAXHOT-PVXXTIHASA-N

Application

Trehalose 6-phosphate (T6P) dipotassium salt has been used as a standard to measure T6P concentrations in potato tubers. It has also been used as a calibration standard in liquid chromatography-mass spectrometry (LC-MS) assay for measurement of T6P in plants.

Biochem/physiol Actions

Trehalose 6-phosphate (T6P) is synthesised from glucose-6-phosphate (G6P) and uridine 5-diphospho-glucose. Trehalose-6-phosphate synthase catalyzes this reaction. T6P serves as a precursor for the synthesis of trehalose. It regulates sugar metabolism in in Arabidopsis plants and promotes its growth. In plants, T6P blocks the action of sucrose nonfermenting-1-related protein kinase 1, which affects the energy and carbon metabolism.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G6526F1502A2013
assay

~95% (TLC)

assay

≥95% anhydrous basis (enzymatic)

assay

≥97% (enzymatic)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

200

form

powder

form

powder

form

amorphous powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

off-white,



color

white

color

white to off-white

color

white to off-white

cation traces

K: 14-17.3%, P: 5.8 - 6.7%

cation traces

K: 10.16-15.88% (ICP anhydrous)

cation traces

K: 20.3-26.2% (dry basis)

cation traces

Na: 13.1-18.5% (dry basis)


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

target_organs

Eyes,Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Trehalose 6-phosphate regulates starch synthesis via posttranslational redox activation of ADP-glucose pyrophosphorylase
Kolbe A, et al.
Proceedings of the National Academy of Sciences of the USA, 102(31), 11118-11123 (2005)
Analysis of low abundant trehalose-6-phosphate and related metabolites in Medicago truncatula by hydrophilic interaction liquid chromatography-triple quadrupole mass spectrometry
Mata AT, et al.
Journal of Chromatography A, 1477(4), 30-38 (2016)
Effects of drought stress and storage on the metabolite and hormone contents of potato tubers expressing the yeast trehalose-6-phosphate synthase 1 gene
Juhasz Z, et al.
Journal of Agricultural Science, 6(5), 142-142 (2014)



Global Trade Item Number

SKUGTIN
T4272-10MG04061832966458
T4272-25MG04061832966465
T4272-100MG04061832966441

Questions

Reviews

No rating value

Active Filters