Skip to Content
MilliporeSigma

Skip To

SML3103

Fluticasone furoate

≥98% (HPLC), powder, glucocorticoid receptor agonist

Synonym(s):

6a,9a-Difluoro-17a-[(2-furanylcarbonyl)oxy]-11β-hydroxy-16a-methyl-3-oxoandrosta-1,4-diene-17β-carbothioic acid S-fluoromethyl ester, GSK 685 698, GSK 685698, GSK685698, GW 685698X, GW685698X, Veramyst, 6α,9α-Difluoro-17α-[(2-furanylcarbonyl)oxy]-11β-hydroxy-6α-methyl-3-oxoandrosta-1,4-diene-17β-carbothioic acid S-fluoromethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H29F3O6S
CAS Number:
Molecular Weight:
538.58
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fluticasone furoate, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

F[C@@]21[C@H]([C@H]4[C@@]([C@@]([C@@H](C4)C)(OC(=O)c5[o]ccc5)C(=O)SCF)(C[C@@H]2O)C)C[C@@H](C3=CC(=O)C=C[C@@]31C)F

InChI

1S/C27H29F3O6S/c1-14-9-16-17-11-19(29)18-10-15(31)6-7-24(18,2)26(17,30)21(32)12-25(16,3)27(14,23(34)37-13-28)36-22(33)20-5-4-8-35-20/h4-8,10,14,16-17,19,21,32H,9,11-13H2,1-3H3/t14-,16+,17+,19+,21+,24+,25+,26+,27+/m1/s1

InChI key

XTULMSXFIHGYFS-VLSRWLAYSA-N

Biochem/physiol Actions

Fluticasone furoate is a highly potent and selective glucocorticoid receptor (GR) agonist (EC50 = 57 pM by A549-based GRE reporter assay; IC50 = 29 nM against 3.2 ng/mL TNF-α-induced NF-κB activation by A549 reporter line; 117 pM against 1 ng/mL LPS-induced TNF-α release from human PBMCs). Fluticasone furoate prevents ovalbumin-induced lung eosinophilia in an allergic lung inflammation model in vivo (30 μg/brown Norway rat intratracheally) with much reduced or no potency toward mineralocorticoid, progesterone, androgen or estrogen receptors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0002274Y0002293Y0002273
assay

≥98% (HPLC)

assay

-

assay

-

assay

-

form

powder

form

-

form

-

form

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Fluticasone furoate


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
SML3103-10MG04065266693263
SML3103-50MG04065266693270

Questions

Reviews

No rating value

Active Filters