Skip to Content
MilliporeSigma

Skip To

SML1877

Tadalafil

≥98% (HPLC), PDE5 inhibitor, powder

Synonym(s):

(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1′,2′:1,6]pyrido[3,4-b]indole-1,4-dione, ICI-351

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H19N3O4
CAS Number:
Molecular Weight:
389.40
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tadalafil, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +68 to +78°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

N21[C@H](Cc3c4c([nH]c3[C@H]2c5cc6c(cc5)OCO6)cccc4)C(=O)N(CC1=O)C

InChI

1S/C22H19N3O4/c1-24-10-19(26)25-16(22(24)27)9-14-13-4-2-3-5-15(13)23-20(14)21(25)12-6-7-17-18(8-12)29-11-28-17/h2-8,16,21,23H,9-11H2,1H3/t16-,21-/m1/s1

InChI key

WOXKDUGGOYFFRN-IIBYNOLFSA-N

Biochem/physiol Actions

Tadalafil is a potent, selective inhibitor of cGMP specific phosphodiesterase type 5 (PDE5) used to treat erectile dysfunction, pulmonary hypertension, and benign prostatic hyperplasia (BPH).
This β-carboline-based drug has a long elimination half-life of about 18 hours.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G9548SML1213F9057
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: ≥10 mg/mL

color

white to beige

color

white to light brown

color

white to beige

color

yellow


Still not finding the right product?

Explore all of our products under Tadalafil


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters