This product should be stored in 2-8°C in powdered format. The solution can be stored at 2-8°C for several weeks or at -20°C for more extended periods.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
Spectinomycin dihydrochloride pentahydrate, powder, BioReagent, suitable for cell culture
biological source
synthetic (inorganic)
Quality Level
product line
BioReagent
form
powder
potency
≥603 μg per mg
storage condition
(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)
technique(s)
cell culture | mammalian: suitable, cell culture | plant: suitable
impurities
endotoxin, tested
color
white to pale buff
solubility
H2O: soluble 50 mg/mL at
antibiotic activity spectrum
Gram-negative bacteria, Gram-positive bacteria
application(s)
agriculture
mode of action
protein synthesis | interferes
storage temp.
2-8°C
SMILES string
O.O.O.O.O.Cl.Cl.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
InChI
1S/C14H24N2O7.2ClH.5H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;;;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H;5*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;;;/m1......./s1
InChI key
DCHJOVNPPSBWHK-UXXUFHFZSA-N
General description
Application
Biochem/physiol Actions
Mode of Resistance: A mutation in rpsE, the gene for ribosomal protein S5, prevents the binding of spectinomycin. Resistance is also conferred by the aminoglycoside-3′-adenyltransferase gene (aadA).
Antimicrobial spectrum: Spectinomycin acts against gram-negative and gram-positive bacteria.
Preparation Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibiotic activity spectrum Gram-negative bacteria, Gram-positive bacteria | antibiotic activity spectrum Gram-negative bacteria, Gram-positive bacteria | antibiotic activity spectrum Gram-negative bacteria, Gram-positive bacteria | antibiotic activity spectrum - |
| technique(s) cell culture | mammalian: suitable, cell culture | plant: suitable | technique(s) - | technique(s) - | technique(s) - |
| mode of action protein synthesis | interferes | mode of action protein synthesis | interferes | mode of action protein synthesis | interferes | mode of action - |
| product line BioReagent | product line - | product line - | product line - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level - |
| application(s) agriculture | application(s) agriculture | application(s) - | application(s) pharmaceutical (small molecule) |
Still not finding the right product?
Explore all of our products under Spectinomycin dihydrochloride pentahydrate
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| S4014-1G | 04061823532228 |
| S4014-25G | 04061835569366 |
| S4014-5G | 04061835522590 |

