Skip to Content
MilliporeSigma

Skip To

PLA0146

Rabbit anti-BRG1/SMARCA4 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATP-dependent helicase SMARCA4, BAF190, BAF190A, BRG1, BRG1-associated factor 190A, BRM/SWI2-related gene 1, CSS4, FLJ39786, MRD16, RTPS2, SNF2, SNF2-BETA, SNF2-like 4, SNF2L4, SNF2LB, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4, SWI2, brahma protein-like 1, global transcription activator homologous sequence, hSNF2b, homeotic gene regulator, mitotic growth and transcription activator, nuclear protein GRB1, protein BRG-1, protein brahma homolog 1, sucrose nonfermenting-like 4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:250- 1:2,000, immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_003063.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... BRG1/SMARCA4(6597)

Immunogen

The epitope recognized by PLA0146 maps to a region between residue 75 and 125 of human BRG1/SMARCA4 using the numbering given in entry NP_003063.2 (GeneID 6597).

Physical form

Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide

Other Notes

The SWI/SNF-related, matrix-associated, actin-dependent regulators of chromatin (SMARC), also called BRG1-associated factors (BAFs), have been identified as components of the human SWI/SNF-like chromatin-remodeling protein complexes. BRG1/SMARCA4 is an ATPase subunit of the SWI/SNF-A chromatin remodeling complex (BAF complex). In BAF complexes, BRG1/SMARCA4 has been shown to interact with BRCA1.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200195HPA064712HPA029981
clone

polyclonal

clone

5B7, monoclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

mouse, monkey, human

species reactivity

human

species reactivity

human

biological source

rabbit

biological source

rat

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

immunohistochemistry: 1:250- 1:2,000, western blot: 1:2,000- 1:10,000, immunoprecipitation (IP): 2-10 μg/mg

technique(s)

ChIP: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: 0.5-1.0 μg/mL using HeLa, HEK-293T, Raji or HepG2 cells extracts

technique(s)

immunofluorescence: 0.25-2 μg/mL

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:500-1:1000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters