Skip to Content
MilliporeSigma

Skip To

P7877

6-Phosphogluconic acid trisodium salt

≥97% (enzymatic)

Synonym(s):

D-Gluconate 6-phosphate trisodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H10Na3O10P
CAS Number:
Molecular Weight:
342.08
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
258-543-6
Beilstein/REAXYS Number:
3814212
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Level

assay

≥97% (enzymatic)

form

powder

impurities

<0.05 mol % glucose-6-phosphate

color

light yellow

solubility

water: 50 mg/mL, clear, colorless

cation traces

Na: 18.8-21.5% (dry basis)

application(s)

agriculture

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H13O10P.3Na/c7-2(1-16-17(13,14)15)3(8)4(9)5(10)6(11)12;;;/h2-5,7-10H,1H2,(H,11,12)(H2,13,14,15);;;/q;3*+1/p-3/t2-,3-,4+,5-;;;/m1.../s1

InChI key

CBQSOSFCUWVWAQ-CXKLNRRHSA-K

Application

6-Phosphogluconic acid has been used in a study to assess a spectrophotometric method for the determination of creative phosphokinase and myokinase. [1] It has also been used in a study to investigate the reversible enzymatic sythesis of diphosphopyridine nucleotide and inorganic pyrophosphate.

Biochem/physiol Actions

Phosphogluconic acid is an intermediate in the pentose phosphate pathway.

Preparation Note

Prepared by a modification of the procedure of Seegmiller, J.E., et al., J. Biol. Chem., 192, 175 (1951).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P6888P8877G6526
biological source

synthetic (organic)

biological source

synthetic (inorganic)

biological source

synthetic (inorganic)

biological source

synthetic (organic)

assay

≥97% (enzymatic)

assay

≥95% anhydrous basis

assay

≥93% dry basis (enzymatic)

assay

≥95% anhydrous basis (enzymatic)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

solubility

H2O: 50 mg/mL, clear, colorless to faintly yellow

solubility

water: 50 mg/mL, clear to hazy, colorless

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Reversible enzymatic sythesis of diphosphopyridine nucleotide and inorganic pyrophosphate
Kornberg, A.
The Journal of Biological Chemistry, 182, 779-793 (1950)
A spectrophotometric method for the determination of creatine phosphokinase and myokinase.
I T OLIVER
The Biochemical journal, 61(1), 116-122 (1955-09-01)
Yasaman Barekatain et al.
Communications biology, 3(1), 328-328 (2020-06-27)
Magnetic resonance (MR) spectroscopy has potential to non-invasively detect metabolites of diagnostic significance for precision oncology. Yet, many metabolites have similar chemical shifts, yielding highly convoluted 1H spectra of intact biological material and limiting diagnostic utility. Here, we show that



Global Trade Item Number

SKUGTIN
P7877-1G04061834398318
P7877-100MG04061835367443
P7877-500MG04061834398325
P7877-5G04061835367450

Questions

  1. What is the solubility of 6-Phosphogluconic acid trisodium salt, with CAS number 53411-70-4? Additionally, is this compound water-soluble?

    1 answer
    1. The product has a water solubility of 50 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters