Skip to Content
MilliporeSigma

Skip To

I8907

Interferon β from rat

≥95%, recombinant, expressed in CHO cells, lyophilized powder, suitable for cell culture

Synonym(s):

IFN-β

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Level

recombinant

expressed in CHO cells

assay

≥95%

form

lyophilized powder

mol wt

~20 kDa

packaging

pkg of 1 vial

concentration

≥100000 units/mL

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... Ifnb1(24481)

Biochem/physiol Actions

Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.
Interferon β belongs to the type 1 interferon family pf proteins.[1] It is also needed for the inhibition of tumor cell growth via cell death, S-phase increase and mediating differentiation.[2] It is also important for bone homeostasis.[3]

Physical form

Lyophilized from phosphate buffered saline (PBS) containing 125 mM trehalose.

Analysis Note

The biological activity is determined in the cytopathic effect inhibition assay using rat epithelial-like (RATEC) cells with vesicular stomatitis virus (VSV). The cytopathic effect of 50% is produced with approximately 1 unit/ml interferon. Since there is no international rat interferon β standard, units are determined with respect to the NIH international standard reference for mouse interferon α/β.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I8657I3275I9032
biological source

rat

biological source

rat

biological source

rat

biological source

mouse

assay

≥95%

assay

≥95%

assay

≥97% (SDS-PAGE)

assay

≥95%

recombinant

expressed in CHO cells

recombinant

expressed in bacteria

recombinant

expressed in E. coli

recombinant

expressed in E. coli

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

buffered aqueous solution


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


C Ren et al.
Gene therapy, 15(21), 1446-1453 (2008-07-04)
Cell-based therapy for cancer is a promising new field. Among cell types that can be used for this purpose, mesenchymal stem cells (MSCs) appear to hold great advantage for reasons including easier propagation in culture, possible genetic modification to express
R B Pepinsky et al.
The Journal of pharmacology and experimental therapeutics, 297(3), 1059-1066 (2001-05-18)
Interferon therapies suffer from a relatively short half-life of the products in circulation. To address this issue we investigated the effects of polyethylene glycol modification (PEGylation) on the pharmacokinetic properties of human interferon (IFN)-beta-1a. PEGylation with a linear 20-kDa PEG
J Adriaansen et al.
Human gene therapy, 17(10), 985-996 (2006-09-21)
Interferon (IFN)-beta has significant immunomodulatory properties and has received much interest as a potentially therapeutic agent for rheumatoid arthritis (RA). Systemic IFN-beta treatment of patients with RA was not effective, probably because of pharmacokinetic issues. Therefore, we studied the effect



Global Trade Item Number

SKUGTIN
I8907-1VL04061833858806

Questions

Reviews

No rating value

Active Filters