Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
206-862-6
MDL number:
Pricing and availability is not currently available.
Product Name
17α-Hydroxypregnenolone,
Quality Segment
SMILES string
C[C@]1(CC[C@H](O)C2)C2=CC[C@]3([H])[C@]1([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(C(C)=O)O
InChI
1S/C21H32O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,15-18,23-24H,5-12H2,1-3H3
InChI key
JERGUCIJOXJXHF-UHFFFAOYSA-N
General description
17α-hydroxypregnenolone is a derived from pregnenolone.
Application
17α-hydroxypregnenolone has been used as a substrate for the enzyme 3β‐hydroxysteroid dehydrogenase (3β‐HSD) expressed in COS1 cells.
Biochem/physiol Actions
17α-hydroxypregnenolone acts as a precursor for cortisol and sex steroids.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| H5002-5G | 04061832850429 |