Skip to Content
MilliporeSigma

Skip To

D9527

Dialysis tubing cellulose membrane

avg. flat width 43 mm (1.7 in.)

Synonym(s):

Cellulose Dialysis Tubing, Cellulose Membrane, Dialysis Membrane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


avg. diam.

27 mm , when full

avg. flat width

43 mm (1.7 in.)

capacity

~175 mL/ft

General description

Typical molecular weight cut-off = 14,000.

Packaging

Supplied in rolls, dry

Preparation Note

Removal of glycerol included as a humectant can be accomplished by washing the tubing in running water for 3-4 hours. Removal of sulfur compounds can be accomplished by treating the tubing with a 0.3% (w/v) solution of sodium sulfide at 80°C for 1 minute. Wash with hot water (60°C) for 2 minutes, followed by acidification with a 0.2% (v/v) solution of sulfuric acid, then rinse with hot water to remove the acid. This tubing will retain most proteins of molecular weight 12,000 or greater.

Analysis Note

May contain glycerin and sulfur compounds in more than trace amounts.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D9277D9402D9652
avg. diam.

27 mm , when full

avg. diam.

6 mm , when full

avg. diam.

49 mm , when full

avg. diam.

21 mm , when full

capacity

~175 mL/ft

capacity

~10 mL/ft

capacity

~640 mL/ft

capacity

~110 mL/ft

avg. flat width

43 mm (1.7 in.)

avg. flat width

10 mm (0.4 in.)

avg. flat width

76 mm (3.0 in.)

avg. flat width

33 mm (1.3 in.)


Still not finding the right product?

Explore all of our products under Dialysis tubing cellulose membrane


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mohammed M Mehanna et al.
Drug development and industrial pharmacy, 46(11), 1787-1799 (2020-09-09)
Staphylococcus aureus is a leading cause of ocular keratitis worldwide, and the upsurge of Methicillin-resistant Staphylococcus Aureus (MRSA) strains necessitated the development of new antimicrobial agents. D-limonene is the major constituent of oil extracted from citrus peel, which has been
Piotr Kołodziejczyk et al.
Journal of food science and technology, 57(1), 222-232 (2020-01-25)
The goal was to assess the effects of partial replacement of wholemeal rye flour with 30%, 40% and 50% of the high-fibre rye flour (HFRF) on nutritional quality and sensory and physicochemical characteristics of breads. The HFRF supplemented breads (SB30
Francisco Rodríguez-Félix et al.
Food science and biotechnology, 29(5), 619-629 (2020-05-19)
The objective of this work was propose antisolvent-dialysis as a new, easy, one-step and reproducible method for obtaining white zein micro- and nanoparticles powder. Firstly, the study by SEM of white zein powder predicted micro- and nanoparticles with spherical morphology



Global Trade Item Number

SKUGTIN
D9527-100FT04061835270033