Skip to Content
MilliporeSigma

Skip To

D7884

Dialysis tubing, benzoylated

Avg. flat width 32 mm (1.27 in.)

Synonym(s):

Dialysis tubing

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


capacity

~100 mL/ft

pore size

2000 NMWCO

storage temp.

2-8°C

Application

Cellulose tubing, benzoylated to reduce pore size. Useful for separating compounds with a molecular weight of ≤1,200 from compounds with a molecular weight >2,000.

Packaging

Supplied in a water pack containing 0.002% 1-hydroxypyridine-2-thione as preservative.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z371106Z371092Z371084
storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

capacity

~100 mL/ft

capacity

-

capacity

-

capacity

-

pore size

2000 NMWCO

pore size

-

pore size

-

pore size

-


Still not finding the right product?

Explore all of our products under Dialysis tubing, benzoylated


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library