Skip to Content
MilliporeSigma

Skip To

D0901

2′-Deoxyguanosine monohydrate

powder, BioReagent, suitable for cell culture, 99-100%

Synonym(s):

9-(2-Deoxy-β-D-ribofuranosyl)guanine, Guanine-2′-deoxyriboside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4 · H2O
CAS Number:
Molecular Weight:
285.26
UNSPSC Code:
12352207
NACRES:
NA.75
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
39814
Assay:
99-100%
Biological source:
synthetic (organic)
Form:
powder
Solubility:
1 M NH4OH: 50 mg/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

product line

BioReagent

assay

99-100%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

1 M NH4OH: 50 mg/mL

SMILES string

O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

InChI

1S/C10H13N5O4.H2O/c11-10-13-8-7(9(18)14-10)12-3-15(8)6-1-4(17)5(2-16)19-6;/h3-6,16-17H,1-2H2,(H3,11,13,14,18);1H2/t4-,5+,6+;/m0./s1

InChI key

LZSCQUCOIRGCEJ-FPKZOZHISA-N

Application

2′-Deoxyguanosine monohydrate has been used in stromal cell purification. It has also been used for the quantification of whole cell dNTP (deoxyribonucleotide triphosphate).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D7145D7400D8668
biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

assay

99-100%

assay

99-100%

assay

≥99%

assay

≥99% (HPLC)

solubility

1 M NH4OH: 50 mg/mL

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

solubility

water: 25 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

H2O: soluble 25 mg/mL, clear to slightly hazy, colorless to faintly yellow

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

cell culture | mammalian: suitable


Still not finding the right product?

Explore all of our products under 2′-Deoxyguanosine monohydrate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters