Skip to Content
MilliporeSigma

Skip To

D0822

DMEM - high glucose

StableCell, with stable glutamine, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

glucose: high
HEPES: no
stable glutamine
sodium pyruvate: yes
phenol red: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

General description

This DMEM-Hi glucose medium differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0697D0819FG0445
sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

components

glucose: high
HEPES: no
stable glutamine
sodium pyruvate: yes
phenol red: yes
NaHCO3: yes

components

glucose: high
sodium pyruvate: yes
stable glutamine: yes
NaHCO3: yes
phenol red: yes
HEPES: yes

components

stable glutamine: yes
phenol red: yes
HEPES: no
sodium pyruvate: no
glucose: high
NaHCO3: yes

components

NaHCO3: yes
stable glutamine: yes
glucose: high
phenol red: yes
HEPES: no
sodium pyruvate: yes

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under StableCell DMEM - high glucose


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sebastien Chaigne et al.
American journal of physiology. Heart and circulatory physiology, 320(3), H1156-H1169 (2021-01-16)
The TRPV4 channel is a calcium-permeable channel (PCa/PNa ∼ 10). Its expression has been reported in ventricular myocytes, where it is involved in several cardiac pathological mechanisms. In this study, we investigated the implication of TRPV4 in ventricular electrical activity.
Adrien Lupieri et al.
Journal of cell science, 133(13) (2020-06-03)
Arterial remodeling in hypertension and intimal hyperplasia involves inflammation and disrupted flow, both of which contribute to smooth muscle cell dedifferentiation and proliferation. In this context, our previous results identified phosphoinositide 3-kinase γ (PI3Kγ) as an essential factor in inflammatory
Burak Gulen et al.
Nature chemistry, 12(8), 732-739 (2020-07-08)
Various pathogenic bacteria use post-translational modifications to manipulate the central components of host cell functions. Many of the enzymes released by these bacteria belong to the large Fic family, which modify targets with nucleotide monophosphates. The lack of a generic



Global Trade Item Number

SKUGTIN
D0822-500ML04061833558157

Questions

Reviews

No rating value

Active Filters