Skip to Content
MilliporeSigma

Skip To

D0431

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

Synonym(s):

3-(N,N-Dimethyldodecylammonio)propanesulfonate, 3-(N,N-Dimethyllaurylammonio)propanesulfonate, 3-(Dodecyldimethylammonio)propanesulfonate, 3-(Lauryldimethylammonio)propanesulfonate, Dodecyldimethyl(3-sulfopropyl)ammonium hydroxide inner salt, Lauryl sulfobetaine, SB3-12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)11N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
335.55
NACRES:
NA.25
PubChem Substance ID:
eCl@ss:
32190102
UNSPSC Code:
12161900
EC Number:
239-002-3
MDL number:
Beilstein/REAXYS Number:
4145308
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

Quality Segment

assay

>99% (TLC)

form

powder

mol wt

micellar avg mol wt 18,500

aggregation number

55

impurities

≤0.0005% Phosphorus (P), ≤0.1% Insoluble matter

ign. residue

≤0.1%

CMC

2-4 mM (20-25°C)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%, sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%, Ca: ≤0.002%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.005%, Mg: ≤0.0005%, NH4+: ≤0.05%, Na: ≤0.005%, Pb: ≤0.001%, Zn: ≤0.0005%

SMILES string

CCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C17H37NO3S/c1-4-5-6-7-8-9-10-11-12-13-15-18(2,3)16-14-17-22(19,20)21/h4-17H2,1-3H3

InChI key

IZWSFJTYBVKZNK-UHFFFAOYSA-N

General description

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate is a zwitterionic surfactant. [1]

Application

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate has been used in a study to assess protein structural change in a mixed system of ionic and zwitterionic surfactants. [2] It has also been used in a study to investigate the interaction between ionic liquid and zwitterionic surfactant.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4023240772T7763
form

powder

form

powder

form

powder

form

-

assay

>99% (TLC)

assay

≥97.0% (dried material, CHN)

assay

≥98.0% (TLC)

assay

≥99% (TLC)

aggregation number

55

aggregation number

55

aggregation number

83

aggregation number

83

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

-

solubility

water: 100 g/L at 20 °C

solubility

H2O: 50 mg/mL

mol wt

micellar avg mol wt 18,500

mol wt

micellar avg mol wt 18,500

mol wt

micellar avg mol wt 30,200

mol wt

micellar avg mol wt 30,200


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Interaction between ionic liquid and zwitterionic surfactant: A comparative study of two ionic liquids with different anions
Behera, K. and S. Pandey
Journal of Colloid and Interface Science, 331, 10-10 (2009)
Yoshiko Moriyama et al.
Journal of oleo science, 60(5), 229-236 (2011-04-20)
The secondary structure of bovine serum albumin (BSA) in the binary surfactant system of anionic sodium dodecyl sulfate (SDS) and zwitterionic N-dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate (DDAPS) was examined at 25°C. The helicity of BSA decreased from 66% to 55% in a solution of
H L Liou et al.
Biochemistry, 40(21), 6475-6485 (2001-05-24)
The tertiary structure of murine adipocyte fatty acid-binding protein (AFABP) is a flattened 10-stranded beta-barrel capped by a helix-turn-helix segment. This helical domain is hypothesized to behave as a "lid" or portal for ligand entry into and exit from the



Global Trade Item Number

SKUGTIN
D0431-5G04061833557679
D0431-25G04061833619384

Questions

Reviews

No rating value

Active Filters