Skip to Content
MilliporeSigma

Skip To

CLS432002

Corning® CoolCell LX Cell Freezing Container

(12 x 1 mL or 2 mL cryogenic vials), green

Synonym(s):

cryovials

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NC.01
UNSPSC Code:
41123300
Manufacturer/tradename:
Corning 432002
Feature:
Holds 12 x 1-2 mL cryovials
Material:
foamed PE
Packaging:
pkg of 1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

foamed PE

feature

Holds 12 x 1-2 mL cryovials

packaging

pkg of 1

manufacturer/tradename

Corning 432002

technique(s)

cryopreservation: suitable

H × diam.

10.11 cm × 11.7 cm , 12.7 mm well diameter

color

green

capacity

(12 x 1 mL or 2 mL cryogenic vials)

General description

Corning CoolCell LX, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The low heat content also ensures that CoolCell LX containers will rapidly return to room temperature when removed from freezer.

  • For 12 standard 1.0 mL to 2.0 mL cryogenic vials.
  • Radially symmetric for uniform vial freezing.
  • Numbered wells for easy sample identification.
  • Beveled lid for secure gripping and easy opening.
  • Exposed vial tops when lid is open for quick, organized removal of frozen samples.

Legal Information

CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS432003CLS432004CLS432005
feature

Holds 12 x 1-2 mL cryovials

feature

Holds 12 x 1-2 mL cryovials

feature

Holds 12 x 1-2 mL cryovials

feature

-

manufacturer/tradename

Corning 432002

manufacturer/tradename

Corning 432003

manufacturer/tradename

Corning 432004

manufacturer/tradename

Corning 432005

material

foamed PE

material

foamed PE

material

foamed PE

material

foamed PE

packaging

pkg of 1

packaging

pkg of 1

packaging

pkg of 1

packaging

pkg of 1

capacity

(12 x 1 mL or 2 mL cryogenic vials)

capacity

(12 x 1 mL or 2 mL cryogenic vials)

capacity

(12 x 1 mL or 2 mL cryogenic vials)

capacity

-

H × diam.

10.11 cm × 11.7 cm , 12.7 mm well diameter

H × diam.

10.11 cm × 11.7 cm , 12.7 mm well diameter

H × diam.

10.11 cm × 11.7 cm , 12.7 mm well diameter

H × diam.

-


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
CLS432002-1EA04061835179510