Skip to Content
MilliporeSigma

Skip To

C5338

Cyanogen bromide-activated-Sepharose 4 Fast Flow

lyophilized powder

Synonym(s):

Cyanogen bromide-activated Agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41106500
NACRES:
NA.56
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

technique(s)

affinity chromatography: suitable

matrix

Sepharose 4 Fast Flow

matrix active group

cyanate or related structures

matrix spacer

1 atom (when ligands are coupled via isourea derivative or related linkage)

swelling

1 g swells to 4-5 mL gel

particle size

45—165 μm

capacity

6-11 μmol/mL coupling capacity (6-AKS)

storage temp.

2-8°C

General description

C5338-10G′s update product number is GE17-0981-01
Cyanogen bromide-activated Sepharose 4 Fast Flow is a resin composed of cross-linked 4% agarose beads that has been pre-activated with cyanogen bromide. Proteins and other molecules containing primary amino groups are coupled directly to the preactivated gel. It allows for multi-point attachment of protein ligands which minimizes ligand leakage. The resin is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.

Application

Cyanogen bromide-activated-Sepharose is used in affinity chromatography and protein chromatography. Cyanogen bromide-activated-Sepharose has been used to discover insecticidal agents, to raise polyclonal antibodies against the yeast invertase glycosyls, and to study immunological processes of xenograft rejection.

Physical form

Lyophilized powder with sugar stabilizers

Legal Information

Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9142H8280E6754
particle size

45—165 μm

particle size

45—165 μm

particle size

-

particle size

45—165 μm

matrix

Sepharose 4 Fast Flow

matrix

Sepharose 4B

matrix

fast-flow highly cross-linked 4% beaded agarose

matrix

Sepharose 6B

matrix active group

cyanate or related structures

matrix active group

cyanate or related structures

matrix active group

N-hydroxysuccinimide active ester

matrix active group

epoxy

capacity

6-11 μmol/mL coupling capacity (6-AKS)

capacity

-

capacity

-

capacity

-

form

lyophilized powder

form

lyophilized powder

form

isopropanol suspension

form

lyophilized powder

technique(s)

affinity chromatography: suitable

technique(s)

affinity chromatography: suitable

technique(s)

protein array: suitable

technique(s)

affinity chromatography: suitable


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Joyce M Bevington et al.
Virology, 357(1), 102-113 (2006-09-09)
Adeno-associated virus (AAV) is a human parvovirus that normally requires a helper virus such as adenovirus (Ad) for replication. The four replication proteins (Rep78, 68, 52 and 40) encoded by AAV are pleiotropic effectors of virus integration, replication, transcription and
M H Zeng et al.
Transplantation proceedings, 38(10), 3312-3314 (2006-12-19)
Human natural killer (NK) cells, which can mediate direct lysis of porcine endothelial cells, play an important role in xenograft rejection. HLA-G, which is a critical molecule in maintaining maternal immune tolerance of semi-allogenic fetus, is able to protect susceptible
Regina Maria Sousa de Araújo et al.
Plant science : an international journal of experimental plant biology, 183, 20-26 (2011-12-27)
Hemagglutinating activity has been associated to presence of lectin, carbohydrate-binding proteins. In this work Crataeva tapia bark lectin (CrataBL) was purified in milligram quantities (28 mg per g of bark) by ion exchange chromatography. The lectin was thermo-stable, ion-independent and



Global Trade Item Number

SKUGTIN
C5338-5G04061833501993
C5338-1G04061833501986
C5338-10G04061833501979

Questions

1–3 of 3 Questions  
  1. How do I store Product C5338, Cyanogen bromide?

    1 answer
    1. This product as it is supplied, is stored at room temperature.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What does Sepharose 4 Fast Flow mean in the description of Product C5338?

    1 answer
    1. Sepharose 4 Fast Flow is a highly crosslinked 4% agarose matrix which gives excellent physical stability and chromatographic qualities.  It was developed to meet high throughput demands of industrial process separations.  The rigidity of the resin permits high flow rates, which in turn gives good resolution in a minimum amount of time

      Helpful?

Reviews

No rating value

Active Filters