Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352207
MDL number:
Pricing and availability is not currently available.
Quality Level
description
for research or for further manufacturing use
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
suitability
endotoxin tested
components
HEPES: yes
L-glutamine: no
NaHCO3: yes
shipped in
ambient
storage temp.
2-8°C
Application
This animal-component free formulation is optimized for high protein production and well as superior expansion of CHO cells.
Other Notes
Proprietary formulation containing inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, recombinant human insulin, plant hydrolysates, other organic compounds, trace elements, and surfactants.
Does not contain antibiotics, antimycotics, L-glutamine, or transferrin. Contains no animal-derived proteins or other components.
Does not contain antibiotics, antimycotics, L-glutamine, or transferrin. Contains no animal-derived proteins or other components.
Legal Information
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C0363-1L | 04061833450512 |
| C0363-50L | 04061837353185 |
| C0363-100L | 04061837353178 |