Skip to Content
MilliporeSigma

Skip To

B3650

Betaine aldehyde chloride

synthetic (organic), ≥99% (TLC), Choline metabolite, powder

Synonym(s):

(Formylmethyl)trimethylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H12ClNO
CAS Number:
Molecular Weight:
137.61
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Betaine aldehyde chloride,

biological source

synthetic (organic)

Quality Level

assay

≥99% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

2-8°C

SMILES string

[Cl-].C[N+](C)(C)CC=O

InChI

1S/C5H12NO.ClH/c1-6(2,3)4-5-7;/h5H,4H2,1-3H3;1H/q+1;/p-1

InChI key

FSKVGWXINKZUKS-UHFFFAOYSA-M

Biochem/physiol Actions

Betaine aldehyde is the physiological intermediate in the oxidation of choline to betaine. Mechanistic studies show that in aqueous solution, it exists predominantly (99%) as the diol (hydrate) form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A5626B6506B3501
biological source

synthetic (organic)

biological source

-

biological source

synthetic (organic)

biological source

synthetic (organic)

assay

≥99% (TLC)

assay

≥99% (TLC)

assay

≥99%

assay

≥99%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder or crystals

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

room temp

solubility

water: 50 mg/mL, clear, colorless to light yellow

solubility

water: 100 mg/mL, clear to slightly hazy, colorless

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: 50 mg/mL, clear, colorless


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Shiro Mitsuya et al.
SpringerPlus, 4, 202-202 (2015-05-21)
Glycinebetaine (GB) is an important compatible solute for salinity tolerance in many plants. In this study, we analyzed the enzymatic activity and the expression level of betaine aldehyde dehydrogenase (BADH), an important enzyme that catalyzes the last step in the
Yongliang Liu et al.
Physiologia plantarum, 163(1), 73-87 (2018-01-04)
As compatible solute, glycine betaine (GB) plays a significant role in salinity tolerance in GB accumulating plants. Solanaceous crops such as tomato (Solanum lycopersicum) and tobacco (Nicotiana tabacum) are salt sensitive and naturally GB non-accumulators. In Solanaceae, only the Lycium
E Ortega et al.
Talanta, 147, 253-260 (2015-11-26)
Choline oxidase (ChOx) is a flavoenzyme catalysing the oxidation of choline (Ch) to betaine aldehyde (BA) and glycine betaine (GB). In this paper a fundamental study of the intrinsic fluorescence properties of ChOx due to Flavin Adenine Dinucleotide (FAD) is



Global Trade Item Number

SKUGTIN
B3650-2MG04061832945354
B3650-100MG04061835553914
B3650-10MG04061832945347

Questions

Reviews

No rating value

Active Filters