Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
Anti-Actin, Cardiac antibody, Mouse monoclonal, clone AC1-20.4.2, purified from hybridoma cell culture
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AC1-20.4.2, monoclonal
form
buffered aqueous solution
mol wt
antigen ~42 kDa
species reactivity
rat, human, bovine, chicken, mouse
concentration
~1.0 mg/mL
technique(s)
immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using digested rat heart tissue, indirect ELISA: suitable, western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ACTC1(70)
mouse ... Actc1(11464)
rat ... Actc1(29275)
General description
Immunogen
Application
Immunohistochemistry (1 paper)
Western Blotting (1 paper)
Physical form
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form ascites fluid | antibody form ascites fluid |
| clone AC1-20.4.2, monoclonal | clone 1A4, monoclonal | clone 5C5, monoclonal | clone AC-40, monoclonal |
| species reactivity rat, human, bovine, chicken, mouse | species reactivity human, mouse, rat, chicken, frog, canine, rabbit, guinea pig, goat, bovine, sheep, snake | species reactivity rabbit, carp, rat, sheep, guinea pig, human, snake, bovine, frog | species reactivity human, Xenopus, hamster, mouse, guinea pig, rat, rabbit, snail, sheep, canine, carp, chicken, pig, bovine, viper, goat |
| Gene Information human ... ACTC1(70) | Gene Information human ... ACTA2(59) | Gene Information human ... ACTA1(58), ACTC1(70) | Gene Information human ... ACTA1(58), ACTA1(58), ACTA1(58), ACTA2(59), ACTA2(59), ACTA2(59), ACTB(60), ACTC1(70), ACTC1(70), ACTC1(70), ACTG1(71), ACTG1(71), ACTG1(71), ACTG2(72), ACTG2(72), ACTG2(72) |
Still not finding the right product?
Explore all of our products under Anti-Actin, Cardiac antibody, Mouse monoclonal
— or —
Try our Product Selector Tool to narrow your options
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
wgk
WGK 1
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A9357-200UL | 04061833402382 |
| A9357-100UL | 04061837718984 |



