Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
goat
Quality Segment
conjugate
agarose conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
suspension
species reactivity
mouse
extent of labeling
≥5 mg per mL
technique(s)
Ouchterlony double diffusion: suitable, immunoprecipitation (IP): suitable
matrix
Cross-linked 4% beaded agarose
matrix activation
cyanogen bromide
matrix spacer
1 atom
capacity
0.4 mg/mL, resin binding capacity (mouse IgG)
storage temp.
2-8°C
target post-translational modification
unmodified
General description
Immunogen
Application
Chromatin immunoprecipitation (1 paper)
Immunoprecipitation (1 paper)
Biochem/physiol Actions
Physical form
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source goat | biological source goat | biological source goat | biological source goat |
| species reactivity mouse | species reactivity - | species reactivity mouse | species reactivity mouse |
| technique(s) Ouchterlony double diffusion: suitable, immunoprecipitation (IP): suitable | technique(s) Ouchterlony double diffusion: suitable | technique(s) Ouchterlony double diffusion: suitable | technique(s) direct ELISA: 1:7,000-1:21,000, western blot: suitable |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form IgG fraction of antiserum | antibody form affinity isolated antibody |
| conjugate agarose conjugate | conjugate unconjugated | conjugate agarose conjugate | conjugate alkaline phosphatase conjugate |
| form suspension | form lyophilized powder | form saline suspension | form buffered aqueous solution |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A6531-2ML | 04061837514517 |
| A6531-5ML | 04061837514524 |
| A6531-25ML | 04061837514500 |




