Skip to Content
MilliporeSigma

Skip To

922P

Trilogy® Pretreatment Solution

greener alternative
Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352203
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For In Vitro Diagnostic Use in Select Regions

Quality Level

form

aqueous solution

packaging

pkg of 1 L ready-to-use (922P-09), pkg of 200 mL concentrated 20x (922P-06), pkg of 200 mL ready-to-use (922P-05), pkg of 240 mL concentrated 100x (922P-07), pkg of 50 mL concentrated 20x (922P-04)

manufacturer/tradename

Cell Marque

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

immunohistochemistry: suitable

greener alternative category

storage temp.

20-25°C

General description

Trilogy® Pretreatment Solution is intended to be used for immunohistochemical assays on formalin-fixed, paraffin-embedded (FFPE) tissue sections. Using this product eliminates the need for multiple solutions to deparaffinize, rehydrate, and antigen retrieve FFPE tissue samples.1 The use of a single solution allows for a standardized pretreatment protocol that produces consistent and reliable results when used in conjunction with Cell Marque Primary Antibodies.

1. Aghassi et al. (2003). Composition and Method for Treating Tissue Samples (U.S. Patent No. 6,649,368). U.S. Patent and Trademark Office. https://rb.gy/gy42ef
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is an environmentally-friendly, non-toxic alternative and eliminates the need for xylene when working with paraffin-embedded tissue, and thus aligns with "Waste Prevention", and ″Designing Safer Chemicals″. Click here for more information.

Physical form

Solution containing pretreatment reagents.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]
Note: This product is similar in use to Trilogy® Pretreatment Solution (920P series products) but does not contain IGEPAL® CA-630 and complies with European Union (EU) Regulation (EC) No 1907/2006, REACH.

Legal Information

IGEPAL is a registered trademark of Solvay
Trilogy is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
920P1.00591D5688
Quality Level

100, 500

Quality Level

500

Quality Level

400

Quality Level

200

technique(s)

immunohistochemistry: suitable

technique(s)

immunohistochemistry: suitable

technique(s)

-

technique(s)

PCR: suitable

form

aqueous solution

form

aqueous solution

form

liquid

form

liquid

manufacturer/tradename

Cell Marque

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative product characteristics

-

greener alternative product characteristics

-

greener alternative category

Aligned

greener alternative category

-

greener alternative category

-

greener alternative category

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

>230.0 °F

flash_point_c

> 110 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Can Trilogy be used in place of ER2 on a BondMax machine?

    1 answer
    1. Unfortunately, it is not possible to make a recommendation in this case. It should be validated the use of any product for their specific needs.

      Helpful?

Reviews

No rating value

Active Filters