Skip to Content
MilliporeSigma

Skip To

911

Naphthol AS-D Chloroacetate Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
205-594-7
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

8 mg/mL

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cc1ccccc1NC(=O)c2cc3ccccc3cc2OC(=O)CCl

InChI

1S/C20H16ClNO3/c1-13-6-2-5-9-17(13)22-20(24)16-10-14-7-3-4-8-15(14)11-18(16)25-19(23)12-21/h2-11H,12H2,1H3,(H,22,24)

InChI key

FMVKYSCWHDVMGO-UHFFFAOYSA-N

Application

For use in Sigma Procedure 91C for the demonstration of specific esterase in blood, bone marrow films, tissue touch preparations, cytocentrifuge preparations and paraffin tissue sections.

Other Notes

Naphthol AS-D chloroacetate, 8 mg/mL, and stabilizer

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8613871916
concentration

8 mg/mL

concentration

4 mg/mL

concentration

12.5 mg/mL

concentration

12.5 mg/mL

form

solution

form

solution

form

solution

form

solution

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Zhibiao Bai et al.
Aging, 12(8), 7534-7548 (2020-04-23)
Intervertebral disc degeneration (IDD) is characterized by the decrease of nucleus pulposus cells (NPCs). With the increase of the degree of degeneration, the reactive oxygen species (ROS) in nucleus pulposus tissue increases. Pyroptosis is a newly discovered form of cell
Shervin Bahrami et al.
Clinical immunology (Orlando, Fla.), 191, 37-43 (2018-03-24)
Retroviruses including Human Endogenous Retroviruses (HERVs), contain a conserved region with highly immunomodulatory functions in the transmembrane proteins in envelope gene (env) named immunosuppressive domain (ISU). In this report, we demonstrate that Env59-GP3 peptide holds therapeutic potential in a mouse



Global Trade Item Number

SKUGTIN
911-10ML04061838203540
911-3ML04061837280085

Questions

Reviews

No rating value

Active Filters