Skip to Content
MilliporeSigma

Skip To

60353

Potassium phosphate dibasic

anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
PubChem Substance ID:
eCl@ss:
38070212
UNSPSC Code:
12161700
NACRES:
NA.25
EC Number:
231-834-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

product line

BioUltra

assay

≥99.0% (T)

form

powder or crystals

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

pH

8.7-9.3 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤10 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤5000 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.20, λ: 280 nm Amax: ≤0.15

suitability

suitable for luminescence

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Application

Potassium phosphate monobasic and dibasic, are used to create phosphate buffered solutions such as potassium phosphate buffered water and saline solutions used in biological applications.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.438397954961.93630
assay

≥99.0% (T)

assay

-

assay

≥98%

assay

-

grade

Molecular Biology

grade

anhydrous

grade

ACS reagent, anhydrous

grade

anhydrous

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

product line

BioUltra

product line

-

product line

Redi-Dri

product line

EMPLURA®

form

powder or crystals

form

solid

form

powder

form

-

pH

8.7-9.3 (25 °C, 1 M in H2O)

pH

9 (20 °C, 10 g/L in H2O)

pH

8.5-9.6 (25 °C, 5% in solution)

pH

-


Still not finding the right product?

Explore all of our products under Potassium phosphate dibasic


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ming Ye et al.
Biosensors & bioelectronics, 73, 208-213 (2015-06-17)
Several volatile organic compounds (VOCs) are released from human breath or skin. Like chemical substances in blood or urine, some of these vapors can provide valuable information regarding the state of the human body. A highly sensitive acetone biochemical gas
Audrey Beaussart et al.
ACS nano, 8(10), 10723-10733 (2014-10-07)
A variety of bacterial pathogens use nanoscale protein fibers called type IV pili to mediate cell adhesion, a primary step leading to infection. Currently, how these nanofibers respond to mechanical stimuli and how this response is used to control adhesion
Ragnar Thomsen et al.
Drug testing and analysis, 7(7), 565-576 (2014-10-28)
Synthetic cannabimimetic agents are a large group of diverse compounds which act as agonists at cannabinoid receptors. Since 2004, synthetic cannabinoids have been used recreationally, although several of the compounds have been shown to cause severe toxicity in humans. In



Global Trade Item Number

SKUGTIN
843872001004027269126339
843872000504027269276102
60353-1KG04061832675701
60353-250G04061832675718

Questions

Reviews

No rating value

Active Filters