Skip to Content
MilliporeSigma

Skip To

08059

Borate Buffered Saline

pH 8.2

Synonym(s):

Saline, Borate Buffered, pH 8.2, tablets

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tablet

Quality Level

pH

8.2

solubility

water: 2 tablet/liter

Application

Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters.

Packaging

100 tablets in blister package

Preparation Note

1 tablet in 500 mL water yields in 0.010 M borate buffer, 0.15 M NaCl, pH 8.2

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5247500798252455
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

tablet

form

-

form

tablet

form

tablet

pH

8.2

pH

-

pH

-

pH

-

solubility

water: 2 tablet/liter

solubility

-

solubility

-

solubility

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Edward Lum et al.
Contact lens & anterior eye : the journal of the British Contact Lens Association, 27(1), 21-26 (2005-11-24)
The study determined the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. One lens type from each FDA contact lens material category was equilibrated to phosphate buffered saline (PBS) or borate buffered saline (BBS), at
Xiu-yan Huang et al.
Zhonghua gan zang bing za zhi = Zhonghua ganzangbing zazhi = Chinese journal of hepatology, 18(12), 915-919 (2011-01-06)
To investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. The siRNA targeting MTSS1 was cloned into one lentivirus work vector.
Alicia Marro et al.
Immunotherapy, 3(8), 971-978 (2011-08-17)
Can heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis be used to treat canine flea allergy? Organisms cultured on Sauton's medium into stationary phase were autoclaved in borate-buffered saline and stored at 10 mg wet weight/ml. Intradermal



Global Trade Item Number

SKUGTIN
A53605-100G04061833375761
A53605-5G04061833375785
A53605-25G04061833375778
08059-100TAB-F04061838648822

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters