Skip to Content
MilliporeSigma

Skip To

STS0216

ECO Brij® S100

greener alternative

average Mn ~4,670

Synonym(s):

Brij® S 100, Polyoxyethylene (100) stearyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C18H37(OCH2CH2)nOH, n~100
CAS Number:
UNSPSC Code:
12165104
NACRES:
NA.28
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Level

form

solid

mol wt

average Mn ~4,670

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1.0% water

mp

51-54 °C (lit.)

acid number

≤2.0 mg KOH/g

hydroxyl value

13‑25 mg KOH/g

density

1.1 g/mL at 25 °C (lit.)

HLB

18.8

greener alternative category

SMILES string

O(CC[O])CCCCCCCCCCCCCCCCCC

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

General description

ECO Brij® S100 is an ethoxylated natural fatty alcohol, a nonionic bio-based surfactant that belongs to the class of polyoxyethylene ethers. This naturally derived detergent offers a number of functional benefits including detergency, wetting, and dispersing making it suitable for a range of applications in biochemical and biological research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Brij is a registered trademark of Croda International PLC
ECO BRIJ is a registered trademark of Croda Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
STS0215388890466387
form

solid

form

solid

form

-

form

-

mol wt

average Mn ~4,670

mol wt

average Mn ~711

mol wt

average Mn ~711

mol wt

average Mn ~4,670

mp

51-54 °C (lit.)

mp

37-39 °C (lit.)

mp

37-39 °C (lit.)

mp

51-54 °C (lit.)

HLB

18.8

HLB

12.4

HLB

12

HLB

18

impurities

≤1.0% water

impurities

-

impurities

-

impurities

-

description

non-ionic

description

non-ionic

description

-

description

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S O Williams et al.
Drug development and industrial pharmacy, 26(1), 71-77 (2000-04-01)
The purpose of this study was to scale up an oil/water (o/w) cream formulation containing 40% diethylene glycol monoethyl ether (DGME), developed via 300-g laboratory batches in a 2(5-2) fractional factorial design, to 7-kg batch sizes in a Brogli-10 homogenizer.
Jae-Haeng Song et al.
Drug development and industrial pharmacy, 35(8), 897-903 (2009-02-26)
The oral administration of loratadine, an antihistamine, can have a variety of adverse side effects, such as headache, fatigue, and nausea, because of the transient high blood concentration. To avoid these effects, loratadine can be administered using a transdermal drug
J Q Zhang et al.
Drug delivery, 14(6), 381-387 (2007-08-19)
The study describes the development of stealth solid lipid nanoparticles (SLNs) as colloidal carriers for silibinin, a drug with very low solubility. Stealth SLNs were constituted mainly of bioacceptable and biodegradable lipids, such as stearic acid and surfactant Brij 78



Global Trade Item Number

SKUGTIN
STS0216-250G04065266169270
STS0216-1KG04065266169263

Questions

Reviews

No rating value

Active Filters