Skip to Content
MilliporeSigma

Skip To

PHR2467

Glycopyrrolate Related Compound C

pharmaceutical secondary standard

Synonym(s):

α-Cyclopentylmandelic acid, Glycopyrronium Bromide Impurity J (PhEur), 2-Cyclopentyl-2-hydroxy-2-phenylacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H16O3
CAS Number:
Molecular Weight:
220.26
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to USP 1296042

API family

glycopyrrolate

form

solid

packaging

pkg of 50 mg

mp

146-151 °C

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

OC(=O)C(O)(C1CCCC1)c2ccccc2

InChI

1S/C13H16O3/c14-12(15)13(16,11-8-4-5-9-11)10-6-2-1-3-7-10/h1-3,6-7,11,16H,4-5,8-9H2,(H,14,15)

InChI key

WFLUEQCOAQCQLP-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2465PHR2468PHR2713
grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

form

solid

form

solid

form

liquid

form

solid

agency

traceable to USP 1296042

agency

traceable to USP 1296029

agency

traceable to USP 1296100

agency

traceable to USP 1296086

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

mp

146-151 °C

mp

-

mp

-

mp

-


Still not finding the right product?

Explore all of our products under Glycopyrrolate Related Compound C


signalword

Danger

Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Corrosion

hcodes

Hazard Classifications

Eye Dam. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
PHR2467-50MG04061842563258

Questions

Reviews

No rating value

Active Filters