Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C4H7N5O · H2SO4 · H2O
CAS Number:
Molecular Weight:
257.23
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
5680241
Pricing and availability is not currently available.
Quality Segment
grade
certified reference material, pharmaceutical secondary standard
API family
folic acid
form
solid
CofA
current certificate can be downloaded
packaging
pkg of 20 mg
mp
≥300 °C
application(s)
pharmaceutical
storage temp.
2-30°C
InChI
1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4)
InChI key
RSKNEEODWFLVFF-UHFFFAOYSA-N
General description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Application
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Analysis Note
These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB4756 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PHR2001-20MG | 04061832567938 |
