Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.24
UNSPSC Code:
12352119
Pricing and availability is not currently available.
grade
certified reference material
Quality Segment
isotopic purity
13C, 18O
manufacturer/tradename
NIST®
format
matrix material
Other Notes
Reference/Informational value is provided for the following analytes.
Carbon (13C/12C), Oxygen (18O/16O)
See certificate for values and more details at nist.gov/SRM.
Carbon (13C/12C), Oxygen (18O/16O)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
1 of 1
This Item | |||
|---|---|---|---|
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename NIST® | manufacturer/tradename NIST® | manufacturer/tradename NIST® | manufacturer/tradename NIST® |
| format matrix material | format matrix material | format matrix material | format mixture |
| isotopic purity 13C, 18O | isotopic purity 13C, 18O | isotopic purity 13C, 18O | isotopic purity - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Press. Gas Liquefied gas
Storage Class
2A - Gases
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| NISTRM8564 | 04061834196976 |


