Skip to Content
MilliporeSigma

Skip To

F0372000

Formoterol fumarate dihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(R*,R*)-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide fumarate, Formoterol hemifumarate salt monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H24N2O4 · 0.5C4H4O4 · H2O
CAS Number:
Molecular Weight:
420.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

formoterol 

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.OC(=O)\C=C\C(O)=O.COc1ccc(C[C@H](C)NC[C@@H](O)c2ccc(O)c(NC=O)c2)cc1.COc3ccc(C[C@H](C)NC[C@@H](O)c4ccc(O)c(NC=O)c4)cc3

InChI

1S/2C19H24N2O4.C4H4O4.2H2O/c2*1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-3(6)1-2-4(7)8;;/h2*3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2H,(H,5,6)(H,7,8);2*1H2/b;;2-1+;;/t2*13-,19+;;;/m00.../s1

InChI key

RATSWNOMCHFQGJ-TUYNVFRMSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Formoterol fumarate dihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

β2-Adrenoreceptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2703F95521286107
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

API family

formoterol 

API family

formoterol 

API family

-

API family

formoterol 


Still not finding the right product?

Explore all of our products under Formoterol fumarate dihydrate


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Repr. 2 - STOT RE 2 - STOT SE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sean R Jesinkey et al.
The Journal of pharmacology and experimental therapeutics, 351(3), 663-673 (2014-10-09)
Skeletal muscle atrophy remains a clinical problem in numerous pathologic conditions. β2-Adrenergic receptor agonists, such as formoterol, can induce mitochondrial biogenesis (MB) to prevent such atrophy. Additionally, atomoxetine, an FDA-approved norepinephrine reuptake inhibitor, was positive in a cellular assay for



Global Trade Item Number

SKUGTIN
F037200004061833611128
F0372000-10MG04061833823804

Questions

Reviews

No rating value

Active Filters