Skip to Content
MilliporeSigma

Skip To

49909

Lanolin

tested according to Ph. Eur.

Synonym(s):

Adeps lanae, Wool fat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352211
EC Number:
232-348-6
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

tested according to Ph. Eur.

autoignition temp.

833 °F

General description

Lanolin is a wool fat.[1] It is a semisolid of high viscosity. Contact allergy to lanolin (wool alcohols), an important sensitizer, in a central London teaching hospital patch-test population has been reported.[2] Simplified extraction of organochlorine and organophosphorus pesticides, frequently found in lanolin, has been proposed.[1] It is utilized as an emulsifier in pharmaceutical and cosmetic formulations.[3]

Application

Lanolin was used to compose the lanolin ointment to estimate the release kinetics of izotretinon and heparin from various semisolid dosage forms by UV-VIS spectroscopy.[4] It was used to prepare the emulsified aqueous solutions containing crystalline dicarboxylic acid. It may be added as excipient to compose a nanoscale-dispersed eye ointment (NDEO) for the treatment of severe evaporative dry eye.[5]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
135688013568711356905
Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

agency

tested according to Ph. Eur.

agency

-

agency

-

agency

-

autoignition temp.

833 °F

autoignition temp.

833 °F

autoignition temp.

-

autoignition temp.

-


Still not finding the right product?

Explore all of our products under Lanolin


Storage Class

11 - Combustible Solids

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters