Skip to Content
MilliporeSigma

Skip To

43925

Nigrosin Stain solution

suitable for microscopy

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
MA.02
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

suitability

suitable for microscopy, Bacillus spp., Corynebacterium spp.

General description

Negative staining method permits visualization of the usually transparent and unstainable capsule of many organisms, most importantly Cryptococcus neoformans.

Other Notes

Composition:
Nigrosin 100 g/l, Formalin 5 ml/l in water

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
944484890045253
technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

suitability

suitable for microscopy, Bacillus spp., Corynebacterium spp.

suitability

-

suitability

-

suitability

suitable for microscopy, Chlamydia spp., Histoplasma spp., Plasmodium spp., Trichomonas spp., bacteria, protozoa, spirochetes

form

liquid

form

liquid

form

-

form

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

application(s)

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

-


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S C Esteves et al.
Fertility and sterility, 66(5), 798-804 (1996-11-01)
To determine whether the hypo-osmotic swelling test can assess viability of frozen spermatozoa. Fresh semen specimens were divided into two aliquots: one was not processed and the other was processed by swim-up. Both aliquots were then cryopreserved by the liquid
Emmanuel I Odongo-Aginya et al.
African health sciences, 7(1), 33-36 (2007-07-03)
The Kato-Katz thick smear technique is the standard technique recommended by the World Health Organisation for the quantitative diagnosis of Schistosoma mansoni and other intestinal helminth infections. The major problem of the technique is that a few hours after the
R R Valle et al.
Theriogenology, 70(1), 115-120 (2008-05-16)
Simple, rapid and stable sperm evaluation methods which have been optimized for common marmoset (Callithrix jacchus) are critical for studies involving collection and evaluation of sperm in the field. This is particularly important for new species groups such as Callitrichidae



Global Trade Item Number

SKUGTIN
695386-25G04061832107424
695386-5G04061832102351
43925-100ML-F04061832246024

Questions

1–2 of 2 Questions  
  1. Does this reagent have an IVD certification compatible with Swiss standards?

    1 answer
    1. This product is a research grade Nigrosin Stain solution that is not IVD certified. Unless otherwise stated, this product is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.

      Helpful?

  2. What's the nigrosin concentration of this product?

    1 answer
    1. The composition of 43925 - Nigrosin Stain solution is as follows:
      Nigrosin 100 g/l, Formalin 5 ml/l in water

      Helpful?

Reviews

No rating value

Active Filters