Skip to Content
MilliporeSigma

Skip To

32471

4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 solution

100 μg/mL in acetone, analytical standard

Synonym(s):

3,6,3-Nonylphenol-13C6, 363-NP-13C6, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-13C6 (ring-13C6)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
13C6C9H24O
CAS Number:
Molecular Weight:
226.31
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-662-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetone

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+6

storage temp.

−20°C

SMILES string

O[13C]1=[13CH][13CH]=[13C](C(CC)(C)CCC(C)C)[13CH]=[13CH]1

InChI

1S/C15H24O/c1-5-15(4,11-10-12(2)3)13-6-8-14(16)9-7-13/h6-9,12,16H,5,10-11H2,1-4H3/i6+1,7+1,8+1,9+1,13+1,14+1

InChI key

BGLUSMSXKMBLFS-FQPQTBQFSA-N

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 solution may be used as a reference standard in the determination of 4-(3,6-dimethyl-3-heptyl)phenol-ring-13C6 solution in environmental samples using gas chromatography coupled with mass spectrometry.

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
335753357433207
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

concentration

100 μg/mL in acetone

concentration

1 μg/mL in acetone

concentration

10 μg/mL in acetone

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mass shift

M+6

mass shift

M+6

mass shift

M+6

mass shift

M+6


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Analysis of Alkylphenols and Alkylphenol Ethoxylates Using new 13C-Labeled and Deuterated Internal Standards According to ISO 18857-2.
Olaf P Heemken and Nicole Amann
Analytix, 7(4), 1-7 (2011)



Global Trade Item Number

SKUGTIN
32471-1ML04061826711675

Questions

Reviews

No rating value

Active Filters