Skip to Content
MilliporeSigma

Skip To

08603

tert-Butyl methyl ether

analytical standard

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
216-653-1
Beilstein/REAXYS Number:
1730942
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

vapor density

3.1 (vs air)

assay

≥99.9% (GC)

autoignition temp.

705 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

15.1 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.369

bp

55-56 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

General description

tert-Butyl methyl ether or methyl tert-butyl ether (MTBE) is an oxygenated compound.. It is commonly used as a gasoline additive in fuel. Though it is known to control air pollution, it is a potential groundwater pollutant.

Application

MTBE may be used as a reference standard for the determination of MTBE in water by headspace-solid-phase microextraction (headspace-SPME) method.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1560179787650560
grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

reagent grade

grade

HPLC Plus

format

neat

format

neat

format

-

format

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

vapor density

3.1 (vs air)

vapor density

3.1 (vs air)

vapor density

3.1 (vs air)

vapor density

3.1 (vs air)

autoignition temp.

705 °F

autoignition temp.

705 °F

autoignition temp.

705 °F

autoignition temp.

705 °F

bp

55-56 °C (lit.)

bp

55-56 °C (lit.)

bp

55-56 °C (lit.)

bp

55-56 °C (lit.)


Still not finding the right product?

Explore all of our products under tert-Butyl methyl ether


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-18.4 °F - closed cup

flash_point_c

-28 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Analysis of methyl tert-butyl ether (MTBE) and tert-butyl alcohol (TBA) in ground and surface water.
Schmidt TC.
TrAC, Trends in Analytical Chemistry, 22(10), 776-784 (2003)
Sensitive method for determination of methyl tert-butyl ether (MTBE) in water by use of headspace-SPME/GC-MS.
Achten C, et al.
Fresenius Journal of Analytical Chemistry, 371(4), 519-525 (2001)
The effects of methyl tert-butyl ether (MTBE) on the male rat reproductive system.
Li D, et al.
Food And Chemical Toxicology, 46(7), 2402-2408 (2008)



Global Trade Item Number

SKUGTIN
557358-1G04061837332692
08603-5ML-F04061838652690

Questions

Reviews

No rating value

Active Filters