Skip to Content
MilliporeSigma

Skip To

04733

Acetyleugenol

analytical standard

Synonym(s):

Eugenyl acetate, 4-Allyl-2-methoxyphenyl acetate, O-Acetyleugenol, Eugenol acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
202-235-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.3% water

refractive index

n20/D 1.517-1.521, n20/D 1.518 (lit.)

bp

281-286 °C (lit.)

density

1.079 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(CC=C)ccc1OC(C)=O

InChI

1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3

InChI key

SCCDQYPEOIRVGX-UHFFFAOYSA-N

General description

Acetyleugenols are p-allylbenzene derivatives of eugenol which are present inherently in traditional foods, specifically in spices like cloves, cinnamon and basil.

Application

Acetyleugenol may be used as a reference standard for the quantification of acetyleugenol in Longhu Rendan pills by gas chromatography coupled with triple quadrupole mass spectrometry (GC-MS/MS), in spice essential oils by gas-liquid chromatography, in the ethanol extracts of Geopropolis from Malaysian stingless bees, Heterotrigona itama by ultra-high-performance liquid chromatography with quadrupole time-of-flight mass spectrometry (UHPLC-Q-TOF/MS) and in oak wood as odorous compounds by two-dimensional GC-MS coupled to olfactometry.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W246905W246700W246719
grade

analytical standard

grade

FG, Fragrance grade, Halal, Kosher

grade

FG, Halal, Kosher

grade

FG, Fragrance grade, Halal, Kosher, natural

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

flavors and fragrances

application(s)

flavors and fragrances

assay

≥98.0% (GC)

assay

≥98%

assay

≥98%

assay

≥98%

format

neat

format

-

format

-

format

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

bp

281-286 °C (lit.)

bp

281-286 °C (lit.)

bp

254 °C (lit.)

bp

254 °C (lit.)


Still not finding the right product?

Explore all of our products under Acetyleugenol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

10 - Combustible liquids

flash_point_f

287.6 °F - closed cup

flash_point_c

142 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Identification of odorous compounds in oak wood using odor extract dilution analysis and two-dimensional gas chromatography-mass spectrometry/olfactometry.
Ghadiriasli, et al.
Analytical and Bioanalytical Chemistry, 410(25), 6595-6607 (2018)
Influence of heating on antioxidant activity and the chemical composition of some spice essential oils.
Tomaino, et al.
Food Chemistry, 89(4), 549-554 (2005)
Rapid Determination of Major Compounds in the Ethanol Extract of Geopropolis from Malaysian Stingless Bees, Heterotrigona itama, by UHPLC-Q-TOF/MS and NMR.
Zhao, et al.
Molecules (Basel), 22(11), 1935-1935 (2017)



Global Trade Item Number

SKUGTIN
191175-1G04061825847603
04733-100MG04061838635761

Questions

Reviews

No rating value

Active Filters