Skip to Content
MilliporeSigma

Skip To

LLDH-RO

Roche

L-Lactate Dehydrogenase (L-LDH)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
~550 units/mg protein (at 25 °C (1,100 U/mg at 37 °C) with pyruvate as the substrate.)
Biological source:
rabbit muscle
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit muscle

form

suspension

specific activity

~550 units/mg protein (at 25 °C (1,100 U/mg at 37 °C) with pyruvate as the substrate.)

mol wt

140,000  Da

packaging

pkg of 10 mL (10127884001 [100 mg]), pkg of 2 mL (10127230001 [10 mg]), pkg of 5 mL (10127876001 [25 mg])

manufacturer/tradename

Roche

technique(s)

activity assay: suitable

color

white

pH

6.0-7.0

solubility

water: miscible

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Aldolase <0.001%, GOT <0.01%, GPT <0.01%, MDH <0.01%, PK <0.001%, myokinase <0.01%

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

L-lactate:NAD+ oxidoreductase.
L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.

Application

Reduction of α-ketoacids to α-hydroxycarboxylic acids, or reverse reaction.[1][2]

Biochem/physiol Actions

L()-Lactate dehydrogenase (LDH) is specific for L()-lactate (relative rate = 100). It does not react with D(-)-lactate.LDH will also slowly oxidize glyoxylate (oxoacetate), glycerate and 3-halogen derivatives of L-lactate (rate with each, ≤ 1); 3-phosphoglycerate does not react.LDH will reduce pyruvate (2-oxopropanate; relative rate = 100) and other 2-oxoacids (e.g., 2-oxobutyrate), with rates decreasing rapidly with increasing chain length of the acid. Heart LDH reacts with 2-oxobutyrate at a greater rate than does skeletal muscle LDH.LDH will also oxidize 2,4-diketoacids (relative rate = 10). The enzyme is realtively specific for NAD(H) (relative rate = 100); NADP(H) is utilized much less efficiently (relative rate < 1). Representative Kms for (hog) skeletal muscle LDH (pH 7.5) are lactate, 8.3 mM; pyruvate, 0.12 mM; NAD, 0.10 mM; NADH, 0.012 mM.Representative Kms for (pig) heart LDH (pH 7.5) are lactate, 3.3 mM; pyruvate, 0.15 mM; NAD, 0.07 mM;NADH, 0.011 mM. In general, the concentration of pyruvate (reduction reaction) or of L-lactate (oxidation reaction) necessary to obtain a maximal rate with LDH is lower for preparations from heart tissue than for the preparations from skeletal muscle.

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 7

Preparation Note

Activator: 2-amino-2-methyl-1-propanol, diethanolamine, fluoride and heparin (oxidation reaction). Sodium sulfite (Na2SO3) will protect the enzyme from the effects of thiol-attacking reagents. Mercaptans can reverse the inhibitory effects of thiol-attacking reagents.

Analysis Note

Absorbance:The purified enzyme from beef heart has an absorbance of 14.9 (10 mg/ml, 280 nm).

Other Notes

For life science research only. Not for use in diagnostic procedures.
LDH is a tetramer (MW = approx. 140,000 D, rabbit muscle or pig heart LDH). In mammals, there are two types of LDH subunits, M and H, with similar molecular weight (subunit MW ≈ 36,500 D) but differing amino acid composition.
Therefore, 5 electrophoretically distinguishable LDH isoenzymes, LDH-1 through LDH-5, of differing subunit composition, are found in mammals.
The molecular weight of all LDH isoenzymes is approximately the same.
– LDH-1 (H4), LDH-2 (H3M): predominant components of heart LDH
– LDH-3 (H2M2): principal component of LDH from lymphatic tissue
– LDH-4 (HM3), LDH-5 (M4): predominate in skeletal muscle or liver LDH

Note: The M subunit of LDH used to be designated the A subunit; the H subunit was the B subunit. Under the old designation, LDH-1 isoenzyme was LDH-B4.
One unit (U) lactate dehydrogenase will reduce 1 µmol of pyruvate to L-lactate in 1 minute at +25 °C and pH 7.0.







Note: For preparations H and I, the activity is defined at +30 °C, pH 7.8. The above assay consumes 1 mol of NADH per mol of pyruvate reduced.





Unit Conversion: For the reduction reaction, pyruvate as substrate:





1 U (+25 °C) 1.5 U (+37 °C) [hog muscle LDH in glycerol].





1 U (+25 °C) 1.8 U (+37 °C) [hog muscle LDH in amm onium sulfate].





1 U (+25 °C) 1.1 U (+30 °C) 2.0 U (+37 °C) [rabbit muscle LDH].





1 U (+25 °C) 1.4 U(+30 °C) 2.5 U (+37 °C) [pig heart LDH].





1 U (+25 °C) 2.5 U (+37 °C) [beef heart LDH].

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
LMDH-RO10107085001427217-M
Gene Information

rabbit ... LOC100355262(100355262)

Gene Information

Porcine ... MDH2(397039)

Gene Information

-

Gene Information

-

technique(s)

activity assay: suitable

technique(s)

activity assay: suitable

technique(s)

-

technique(s)

-

specific activity

~550 units/mg protein (at 25 °C (1,100 U/mg at 37 °C) with pyruvate as the substrate.)

specific activity

~1200 units/mg protein (At 25 °C with oxaloacetate as the substrate.)

specific activity

~550 units/mg protein (at 25 °C (1,000 U/mg at 37 °C) with pyruvate as the substrate.)

specific activity

≥200 units/mg protein

biological source

rabbit muscle

biological source

Porcine heart (mitochondrial)

biological source

hog muscle

biological source

-

form

suspension

form

solution, suspension

form

suspension

form

lyophilized

application(s)

life science and biopharma

application(s)

life science and biopharma

application(s)

-

application(s)

-


Still not finding the right product?

Explore all of our products under L-Lactate Dehydrogenase (L-LDH)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
1012787600104061838667540
1012723000104061838667243
1012788400104061838667564