Skip to Content
MilliporeSigma

Skip To

SCT134

BioTracker 510 Green C18(FM3-25) Synaptic Dye

Live cell imaging synaptic dye that detects neural synaptic activitiy at neuromuscular junctions and synapses.

Synonym(s):

Live cell imaging probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

2-8°C

General description

BioTracker synaptic dyes are a series of fluorescent cationic styryl dyes developed to follow synaptic activities at neuromuscular junctions or synapses. These dyes typically have a lipophilic tail at one end and a highly hydrophilic at the other end. In the presence of cells or tissue preparations, the dyes partition between the aqueous phase, where the dyes are virtually non-fluorescent, and the outer leaflet of the cell surface membranes, where the dyes insert the lipophilic end into the membranes and become intensely fluorescent. During endocytosis following nerve stimulation, the dyes become trapped inside the vesicles. Thus, after washing off the dyes on the cell surface, the fluorescent signal is proportional to the number of newly formed vesicles. On the other hand, during exocytosis, the dyes are released from the vesicles along with neurotransmitters, causing a decrease in fluorescent signal. As a result, the change in fluorescent intensity reflects the amount of endocytosis/exocytosis or synaptic activity.

BioTracker 510 Green C18(FM3-25) Synaptic Dye has the longest lipophilic tail (C18). Because of its large size, it is used as a control for ion channel permeability of nerve terminal dyes.

Spectral Properties
Absorbance (MeOH): 510 nm
Emission (MeOH): 625 nm

Absorbance (Membranes): 480 nm
Emission (Membranes): 600 nm

Application

Live cell fluorescent imaging
Live cell imaging synaptic dye that detects neural synaptic activitiy at neuromuscular junctions and synapses.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Preparation Note

Store BioTracker 510 Green C18(FM3-25) Synaptic Dye at 2-8°C, protected from light. Stock solutions can be prepared at 10 mM and stored at 4°C or -20°C for six months or longer.

Analysis Note


Spectral Properties
Absorbance (MeOH): 510 nm
Emission (MeOH): 625 nm

Absorbance (Membranes): 480 nm
Emission (Membranes): 600 nm

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCT133SCT126SCT131
technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters