Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352207
NACRES:
NA.47
eCl@ss:
32161000
Pricing and availability is not currently available.
technique(s)
cell based assay: suitable
detection method
fluorometric
shipped in
ambient
storage temp.
-10 to -25°C
General description
Hypoxia is defined as the reduction or lack of oxygen in organs, tissues, or cells. Hypoxia in living tissue closely relates to a wide array of pathologies, including cancer and ischemia. Existing hypoxia detection probes, such as pimonidazole, require cell fixation followed by immunostaining.
The BioTracker 520 Green Hypoxia Dye is a fluorescent imaging probe for the detection of hypoxia in living cells. This probe is as sensitive as pimonidazole and can be used for live cell fluorescence imaging and flow cytometry applications.
Spectral Properties
Absorbance: 498nm
Emission: 520nm
The BioTracker 520 Green Hypoxia Dye is a fluorescent imaging probe for the detection of hypoxia in living cells. This probe is as sensitive as pimonidazole and can be used for live cell fluorescence imaging and flow cytometry applications.
Spectral Properties
Absorbance: 498nm
Emission: 520nm
Application
Live cell fluorescent imaging
Research Category
Cell Imaging
Cell Imaging
Research Sub Category
Live Cell Dye
Live Cell Dye
Physical form
Lyophilized
Preparation Note
Store BioTracker 520 Green Hypoxia Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) cell based assay: suitable |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
| storage temp. -10 to -25°C | storage temp. -10 to -25°C | storage temp. -10 to -25°C | storage temp. -10 to -25°C |
| detection method fluorometric | detection method fluorometric | detection method fluorometric | detection method fluorometric |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SCT033 | 04054839545665 |



