Skip to Content
MilliporeSigma

Skip To

ABN899

Anti-Olig-2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Oligodendrocyte transcription factor 2, bHLHe19, Olg-2, Oligo2, RACK17, RK17

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, IP, WB
Citations:
5
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OLIG2(10215)

General description

Oligodendrocyte transcription factor 2 (UniProt Q9EQW6; also known as bHLHe19, Olg-2, Oligo2, RACK17, RK17) is encoded by the Olig2 (also known as Bhlhb1) gene (Gene ID 50913) in murine species. Olig2 is a basic helix-loop-helix (bHLH) transcription factor expressed in a spinal cord ventricular zone domain where it mediates the sequential generation of motoneurons and oligodendrocytes during embryogenesis. Olig2 also functions as an anti-neurogenic factor in pMN progenitors at early time points to sustain the cycling progenitor pool. The anti-neurogenicity of Olig2 is also responsible for its role in tumorigenesis, especially in brain tumors and leukemia. Due to its expression in glioblastoma and other diffuse gliomas, Olig2 is a useful diagnostic marker of these brain tumors.
~37 kDa observed. 32.40 kDa calculated.

Immunogen

KLH-conjugated linear peptide corresponding to a sequence in the N-terminal region of mouse Olig-2.

Application

Anti-Olig-2 Antibody is an antibody against Olig-2 for use in Western Blotting, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin).
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated Olig-2 in 500 µg of mouse brain tissue lysate.
Immunohistochemistry Analysis: An 1:500 dilution from a representative lot detected Olig-2 in rat hindbrain tissue sections.
Immunocytochemistry Analysis: An 1:500 dilution from a representative lot detected Olig-2 in PC12 cells.

Physical form

Purified rabbit polyclonal antibody in PBS and 0.1 % sodium azide.

Analysis Note

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Olig-2 in 10 µg of mouse brain tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: AB15328

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB9610AB15328AV31464
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

unconjugated

species reactivity

mouse, rat, human

species reactivity

mouse, human, rat

species reactivity

rat

species reactivity

rat, human, mouse, dog, guinea pig, horse

UniProt accession no.

Q9EQW6

UniProt accession no.

Q13516

UniProt accession no.

Q13516

UniProt accession no.

Q13516


Still not finding the right product?

Explore all of our products under Anti-Olig-2 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters