Skip to Content
MilliporeSigma

Skip To

8.22286

Sodium sulfate

anhydrous for synthesis

Synonym(s):

Sulfuric acid sodium salt (1:2)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
NACRES:
NA.22
EC Index Number:
231-820-9
UNSPSC Code:
12352302
MDL number:
Grade:
synthesis grade
Form:
crystals
Solubility:
200 g/L
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Level

form

crystals

potency

>2000 mg/kg LD50, oral (Rat)

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

bulk density

1400‑1600 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Application

Sodium sulfate (anhydrous) can be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.066491.9363571963
form

crystals

form

solid

form

-

form

crystalline

solubility

200 g/L

solubility

-

solubility

-

solubility

water: soluble 185 g/L

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

100

grade

synthesis grade

grade

ACS reagent

grade

anhydrous

grade

anhydrous

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

pH

-

pH

-


Still not finding the right product?

Explore all of our products under Sodium sulfate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Syntheses of tris (pyrazolyl) methane ligands and {[tris (pyrazolyl) methane] Mn (CO)3}SO3CF3 complexes: Comparison of ligand donor properties
Reger DL, et al.
Journal of Organometallic Chemistry, 607(1-2), 120-128 (2000)
Frank J Schenck et al.
Journal of AOAC International, 85(5), 1177-1180 (2002-10-11)
Water-miscible solvents, such as acetone and acetonitrile, effectively extract both polar and nonpolar pesticide residues from nonfatty foods. The addition of sodium chloride to the resulting acetonitrile-water or acetone-water extract (salting out) results in the separation of the water from



Global Trade Item Number

SKUGTIN
822286500004022536487786
822286100004022536487779

Questions

Reviews

No rating value

Active Filters