Skip to Content
MilliporeSigma

Skip To

8.17073

Fumaric acid

EMPROVE® ESSENTIAL, ChP, NF, JPE

Synonym(s):

Fumaric acid, trans-1,2-Ethylentricarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H4O4
CAS Number:
Molecular Weight:
116.07
UNSPSC Code:
12352106
EC Index Number:
203-743-0
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ChP, JPE, NF

vapor pressure

<0.001 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

375 °C

potency

9300 mg/kg LD50, oral (Rat), 20000 mg/kg LD50, skin (Rabbit)

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

pH

2.1 (20 °C, 4.9 g/L in H2O)

bp

290 °C/1013 hPa (sublimed)

mp

287 °C

transition temp

flash point 273 °C

solubility

4.9 g/L

density

1.64 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[O-]C(=O)\C=C\C(=O)[O-].[H+].[H+]

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Fumaric acid is a commonly used ingredient in both food and pharmaceutical applications. In pharmaceutical applications, fumaric acid has been used primarily as a pH modifier.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1270128670876635
application(s)

liquid formulation
pharmaceutical
solid formulation

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

agency

ChP, NF, JPE

agency

traceable to Ph. Eur. F0600000, traceable to USP 1286708

agency

-

agency

-

form

solid

form

-

form

-

form

-

density

1.64 g/cm3 at 20 °C

density

-

density

-

density

-

solubility

4.9 g/L

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Fumaric acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

523.4 °F

flash_point_c

273 °C



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
817073905004022536931814
817073100004022536702599

Questions

Reviews

No rating value

Active Filters