Skip to Content
MilliporeSigma

Skip To

8.02301

1,1′-Carbonyldiimidazole

greener alternative

for synthesis

Synonym(s):

1,1′-Carbonyldiimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H6N4O
CAS Number:
Molecular Weight:
162.15
UNSPSC Code:
12352115
EC Index Number:
208-488-9
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (GC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (GC)

form

powder

potency

1071 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Carbonylations

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

7.7 (20 °C, 50 g/L in H2O)

mp

116-120 °C

application(s)

peptide synthesis

greener alternative category

storage temp.

2-8°C

SMILES string

[n]2(cncc2)C(=O)[n]1cncc1

InChI

1S/C7H6N4O/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6H

InChI key

PFKFTWBEEFSNDU-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. 1,1′-carbonyldiimidazole (CDI) is an alternative to highly toxic insidious poison phosgene and thus aligns with the principle "Designing Safer Chemicals". Click here for more information.

Application


Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 115 °C
Melting range (upper value): ≤ 121 °C
Identity (IR): passes test


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

8A - Combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802301000204027269276522
802301002504022536372655
802301010004022536372662
802301000504022536372648
802301250004027269678319
802301999904022536372679

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters