Skip to Content
MilliporeSigma

Skip To

494128

Ochratoxin A

≥97% (HPLC), powder, mycotoxin, Calbiochem

Synonym(s):

Ochratoxin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H18ClNO6
CAS Number:
Molecular Weight:
403.81
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ochratoxin A, Ochratoxin A (OTA) is a mycotoxin that acts through mechanisms such as the inhibition of protein synthesis, and may also be mediated by organic anion transporters (OATs).

Quality Segment

assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL, nearly colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c2c(c(c(c1)C(=O)N[C@@H](Cc3ccccc3)C(=O)O)O)C(=O)O[C@@H](C2)C

InChI

1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15+/m1/s1

InChI key

RWQKHEORZBHNRI-BMIGLBTASA-N

General description

Ochratoxin A (OTA) is a mycotoxin that acts through mechanisms such as the inhibition of protein synthesis, and may also be mediated by organic anion transporters (OATs).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Anzai, N., et al. 2010. Toxins2, 1381.
Chopra, M., et al. 2010. Cell Biol. Toxicol.26, 239.
Luhe, A., et al. 2002. Tox. Sci.73, 315.
Dirheimer, G., et al. 1991. IARC Swci Publ.115, 171.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O1877CRM46912O3007
assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

-

assay

-

form

powder

form

powder

form

-

form

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

solubility

DMSO: 50 mg/mL, nearly colorless

solubility

ethanol: soluble

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Ochratoxin A


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
494128-1MG04055977272864

Questions

Reviews

No rating value

Active Filters