Skip to Content
MilliporeSigma

Skip To

475989

MTT

Membrane-permeable yellow dye that is reduced by mitochondrial reductases in living cells to form the dark blue product, MTT-formazan.

Synonym(s):

MTT, 3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium Bromide, Thiazole Blue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H16BrN5S
CAS Number:
Molecular Weight:
414.32
UNSPSC Code:
12171500
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

description

RTECS - XF8060000

assay

≥98% (silver nitrate titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.5% Sulfated ash

color

yellow

solubility

PBS: 2 mg/mL

cation traces

heavy metals: ≤0.001%

shipped in

ambient

storage temp.

15-25°C

SMILES string

[s]1c(nc(c1C)C)[n]2[n+](nc(n2)c4ccccc4)c3ccccc3.[Br-]

InChI

1S/C18H16N5S.BrH/c1-13-14(2)24-18(19-13)23-21-17(15-9-5-3-6-10-15)20-22(23)16-11-7-4-8-12-16;/h3-12H,1-2H3;1H/q+1;/p-1

InChI key

AZKSAVLVSZKNRD-UHFFFAOYSA-M

General description

Membrane-permeable yellow dye that is reduced by mitochondrial reductases in living cells to form the dark blue product, MTT-formazan. Spectrophotometric measurement of MTT-formazan at 540 or 570 nm allows quantitation of cell viability. Used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. An electron acceptor used for studying NADP-dependent dehydrogenases.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Used as a direct indicator of cytotoxicity and apoptosis.
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C.). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Liu, Y., et al. 1997. J. Neurochem.69, 581.
Abdallah, M.A. 1980. Eur. J. Biochem. 112, 331.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5655M212811585029001
color

yellow

color

-

color

dark yellow

color

-

form

solid

form

powder

form

powder

form

crystals

solubility

PBS: 2 mg/mL

solubility

H2O: 5 mg/mL

solubility

2-methoxyethanol: soluble 20 mg/mL, H2O: 5 mg/mL

solubility

-

assay

≥98% (silver nitrate titration)

assay

≥97.5% (HPLC)

assay

98%

assay

-

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jong-Chan Park et al.
Nature communications, 12(1), 280-280 (2021-01-14)
Developing effective drugs for Alzheimer's disease (AD), the most common cause of dementia, has been difficult because of complicated pathogenesis. Here, we report an efficient, network-based drug-screening platform developed by integrating mathematical modeling and the pathological features of AD with
Yi Shan Huang et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 7202519-7202519 (2020-05-19)
Hypertensive renal injury is a primary etiology of end-stage renal disease, and satisfactory therapeutic strategies are urgently required. Cordyceps cicadae, a traditional Chinese herb, has potential renoprotective benefits and is widely used in the treatment of many kidney diseases. To
Ning Wang et al.
Journal of inorganic and organometallic polymers and materials, 1-15 (2020-05-13)
Vulnerability of respiratory mucosa to invasions of airborne pathogens, such as SARS-CoV, MERS-CoV and avian viruses which sometimes cause a life-threatening epidemic and even pandemic, underscores significance of developing a pulmonary vaccine adjuvant-delivery system (VADS). Herein, 30-nm aluminum nanoparticles (ANs)



Global Trade Item Number

SKUGTIN
475989-1GM07790788050603
475989-10GM07790788050597

Questions

Reviews

No rating value

Active Filters