Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
4-Methylumbelliferyl-6-sulfo-N-acetyl-β-D-glucosaminide, Potassium Salt, Fluorogenic substrate for isoenzyme A of N-acetyl-β-D-glucosaminidase (hexosaminidase A). Can be used for the prenatal diagnosis of Tay-Sachs disease and for the classification of GM2 gangliosidosis genotypes.
Quality Level
assay
≥98% (HPLC)
form
crystalline solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated, protect from light
color
white
solubility
water: 20 mg/mL
fluorescence
λex 360 nm, λem 440 nm
shipped in
ambient
storage temp.
−20°C
General description
Preparation Note
Other Notes
Bayleran, J., et al. 1984. Clin. Chim. Acta143, 73.
Inui, K., and Wenger, D.A. 1984. Clin. Genet.25, 93.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| form crystalline solid | form solid | form crystalline solid | form solid |
| assay ≥98% (HPLC) | assay - | assay ≥98% (TLC) | assay ≥99% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
| storage temp. −20°C | storage temp. 10-30°C | storage temp. −20°C | storage temp. 10-30°C |
| storage condition OK to freeze, protect from light, desiccated | storage condition OK to freeze, desiccated, protect from light | storage condition OK to freeze, desiccated, protect from light | storage condition OK to freeze, desiccated, protect from light |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 454428-25MG | 04055977204117 |



