Skip to Content
MilliporeSigma

Skip To

361507

Glycogen, Molecular Biology Grade, Mussel

Suitable as a carrier substance for the precipitation of nucleic acids. One µl is sufficient to precipitate picogram quantities of nucleic acid with Ethanol.

Synonym(s):

Glycogen, Molecular Biology Grade, Mussel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

description

Merck USA index - 14, 4496

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

foreign activity

nucleases, nucleic acids, proteases, none detected

shipped in

wet ice

storage temp.

−20°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-20(45-23-17(36)12(31)10(29)6(2-26)42-23)14(33)18(37)24(43-8)44-19-7(3-27)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21+,22+,23-,24-/m1/s1

InChI key

BYSGBSNPRWKUQH-UJDJLXLFSA-N

General description

One µl is sufficient to precipitate picogram quantities of nucleic acid with Ethanol.
Suitable as a carrier substance for the precipitation of nucleic acids. One µl is sufficient to precipitate picogram quantities of nucleic acid with Ethanol.

Application


  • Nucleic acid contamination of glycogen used in nucleic acid precipitation and assessment of linear polyacrylamide as an alternative co-precipitant.: This study investigates the nucleic acid contamination of glycogen used in nucleic acid precipitation processes and explores linear polyacrylamide as a potential alternative co-precipitant. The research highlights significant implications for biochemical protocols, emphasizing the need for contaminant-free reagents in molecular biology applications. (Bartram et al., 2009).

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G1767G1508G8751
grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

color

clear colorless

color

-

color

white to off-white

color

white to off-white

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
361507-1ML07790788049577

Questions

Reviews

No rating value

Active Filters