Skip to Content
MilliporeSigma

Skip To

229101

Cholic Acid, Sodium Salt

Primary bile acid.

Synonym(s):

Cholic Acid, Sodium Salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H39O5 · Na
CAS Number:
Molecular Weight:
430.55
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25
Assay:
>96% (TLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 2203

Quality Segment

assay

>96% (TLC)

form

solid

mol wt

micellar wt 900

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

pKa 

5.2

CMC

9 - 15 mM

solubility

acetic acid: 1 mg/mL, ethanol: 1 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@H]3O)C[C@@H](CC4)O)C)C[C@@H]2O)CC1)C)C

InChI

1S/C24H40O5.Na/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29);/q;+1/p-1/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-;/m1./s1

InChI key

NRHMKIHPTBHXPF-TUJRSCDTSA-M

General description

Primary bile acid.

Application


  • EXPLOITING THE VERSATILITY OF CHOLESTEROL IN NANOPARTICLES FORMULATION.: This study explores the utility of cholic acid sodium salt in formulating nanoparticles with enhanced functionality for targeted drug delivery systems (Ruozi et al., 2016).

  • Comparative study on the enantiomer separation of 1,1′-binaphthyl-2,2′diyl hydrogenphosphate and 1,1′-bi-2-naphthol by liquid chromatography and capillary electrophoresis using single and combined chiral selector systems.: Explores the role of cholic acid sodium salt in improving the efficiency of chiral separation techniques, vital for the development of pharmaceutical agents (Sybilska et al., 2002).

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
220411T4009360512
assay

>96% (TLC)

assay

>95% (TLC)

assay

≥95% (HPLC)

assay

100.1% dry basis (titration)

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

solubility

acetic acid: 1 mg/mL, ethanol: 1 mg/mL

solubility

water: 20 mg/mL, aqueous buffer: soluble

solubility

-

solubility

water: 200 mg/mL, ethanol: soluble

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

room temp

storage temp.

15-25°C

form

solid

form

solid

form

powder

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
229101-50GM04055977218190
229101-250GM04055977218183

Questions

Reviews

No rating value

Active Filters