Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥95% (SDS-PAGE)
form
liquid
specific activity
≥2000 C5b,6H50 units/mg
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Packaging
Other Notes
Lachmann, P.J. and Thompson, R.A. 1970. J. Exp. Med. 131, 643.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥90% (SDS-PAGE) |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 204906-50UG | 04055977219883 |



